Hand Evaluation Set, Three Pieces, AMERICAN 3B SCIENTIFIC SE
# de catalogue 470119-594
Fournisseur: American 3B Scientific
Spécifications
- Description:Hand Evaluation Set, 3 pieces
- Cat. no.:470119-594
- Supplier No.:W50174
Spécifications
A propos de cet article
The baseline hydraulic hand evaluation set has all instrument necessary to evaluate strength and range of motion of hand and fingers. The hydraulic system assures convenience, product reliability, measurement accuracy and repeatability.
- Standard head hand dynamometer is used by therapists
- The standard 6.35 cm (2.5") diameter head is used throughout the industry as most popular size
Delivery: Includes a 200 lbs. (90 kg) hydraulic hand dynamometer, 50 lbs. (22.6 kg) hydraulic pinch gauge and stainless steel finger goniometer.
Customers Who Bought This Also Bought