Baseline® Hand Evaluation Set, Seven Pieces
# de catalogue 470119-592
Fournisseur: American 3B Scientific
Spécifications
- Description:Hand Evaluation Set
- Cat. no.:470119-592
- Supplier No.:W50173
Spécifications
A propos de cet article
A full analog evaluation set.
- Seven piece evaluation set
- Includes case for storage and transport
The Baseline hand evaluation set includes all instruments needed for a full hand evaluation.
Grip strength is measured by the 200 lbs. hydraulic hand dynamometer and pinch strength by the mechanical 30 lbs. pinch gauge.
Delivery: Includes a stainless steel finger goniometer 5 cm (2"), two-point discriminator with third point, Wartenburg pinwheel, finger circumference gauge, and functional finger motion gauge in a hard shell carrying case.
Customers Who Bought This Also Bought