Back-Leg-Chest Dynamometer, AMERICAN 3B SCIENTIFIC SE
# de catalogue 470119-590
Fournisseur: American 3B Scientific
Spécifications
- Description:Back-Leg-Chest Dynamometer Regular Base
- Cat. no.:470119-590
- Supplier No.:W50172
Spécifications
A propos de cet article
Back-leg-chest dynamometer's to measure the strength of a patients back, leg or chest. Oversized body includes a solid base for safety, as well as cushion handgrips for comfort. Chain adjusts for height differences or to vary the point of force application.
Strength indicator remains at subject's maximum reading until reset.
Scale measure to 300 kg (660 lbs.).
Customers Who Bought This Also Bought