Digital Inclinometer
# de catalogue 470119-586
Fournisseur: American 3B Scientific
Spécifications
- Description:Digital Inclinometer
- Cat. no.:470119-586
- Supplier No.:W50170
Spécifications
A propos de cet article
Motion in its total range can be read from the LED panel, after the joint has been taken through the digital inclinometer. Place the inclinometer near the joint to be measured.
Press "zero" button. Move joint through its range. "HOLD" button.
Includes carrying case.
Customers Who Bought This Also Bought