ContactUsLinkComponent
HBc/HIV/HCV Antibody Test Controls
HBc/HIV/HCV Antibody Test Controls
  10769-911
 :  MEDMIRA
 :  
Test Control
HBc/HIV/HCV Antibody Test Controls
  10769-911
 :  MEDMIRA
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
  • Description:
    HBc/HIV/HCV Antibody Test Controls
  • Cat. no.:
    10769-911
  • Supplier No.:
    815311006104
HBc/HIV/HCV Antibody Test Controls (Test Controls) are external quality control agents, intended for use with the following Miriad rapid antibody tests:
Model 815311006012 Miriad Rapid HCV/HIV Antibody Test POU+
Model 815311005954 Miriad Rapid HCV/HIV Antibody Test LAB+
Model 815311006005 Miriad Rapid HBc/HIV/HCV Antibody Test POU+
Model 815311005978 Miriad Rapid HBc/HIV/HCV Antibody Test LAB+
Performance may vary if used with other rapid tests.
Ordering information: Prior to reconstitution, test controls can be stored at 2 to 30 °C (35 to 85 °F) for up to 18 months. After reconstitution, the test controls are stable for up to 21 days when stored at 2 to 8 °C (35 to 45 °F). Each test control is sufficient for performing 5 tests.
Delivery: Each package includes 1 HBc Positive Test Control, 1 HIV Positive Test Control, 1 HCV Positive Test Control, 1 HBc/HIV/HCV Negative Test Control, 1 vial of reconstitution buffer, 20 transfer pipettes and a package insert.
Customers Who Bought This Also Bought