Corning® Multicoated Microplates, Corning
Fournisseur: Corning
Certificats
A propos de cet article
The Corning® multicoated microplate provides users access to six different surface treatments on a single plate.
This microplate is useful when determining the correct surface for assay requirements. Single surface microplates can then be used for the full screen or experiment. These multicoated plates are currently available in 96-well, 384-well, and 1536-well formats in black with clear bottom.
Customers Who Bought This Also Bought