Corning®, Microplates, High Content Imaging Glass Bottom, Corning
Supplier: Corning
Certificates
About this item
High optical quality glass bottom, black microplates are ideal for performing high content cell based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth.
- High optical quality and scratch-resistant glass
- Glass bottom thickness of 200 µm is well suited for imaging microscopy
- Well bottom flatness <50 µm to ensure planarity for imaging devices
- Low background fluorescence and minimal crosstalk to provide the highest possible optical quality for cell based assays
- Half area 96 well microplate reduces reagent consumption
- 384 well microplate geometry eliminates reagent loss due to 'wicking'
Specifications
- Catalog No:
- 30621-096
- 89511-320
- 89511-324
- 89511-328
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- I-RECs, ISO 14001 - Bedford
- I-RECs, ISO 14001 - Bedford
- I-RECs, ISO 14001 - Bedford