Accessories for U-2D™ 96-Well Plate Sample Control System, MicroSolv
Fournisseur: MicroSolv Technology Corp
Certificats
Accessories for U-2D™ 96-Well Plate Sample Control System, MicroSolv
Fournisseur: MicroSolv Technology Corp
A propos de cet article
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
73,75 $ to 568,62 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
86,08 $ to 717,60 $
WHEATON® MicroLiter Crimp-Top Headspace Vials, 20 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
109,95 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
118,61 $ to 1 128,11 $
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
438,81 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
43,20 $ to 44,79 $