Disposable Carbon Steel Surgical Blades, OR Grade, Sklar®
Supplier: Sklar
Certificates
About this item
Disposable Surgical Blades are carbon steel, industry standard
Commonly used for large, straight incisions. Blades feature a curved cutting edge. Supplied sterile in a box of 100.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.