ContactUsLinkComponent
Pinard Fetoscope, Sklar
Pinard Fetoscope, Sklar
# de catalogue 10194-492
Fournisseur:  Sklar
Fetoscope
Pinard Fetoscope, Sklar
# de catalogue 10194-492
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Silver
  • Cat. no.:
    10194-492
  • Length:
    20.3 cm (8")
  • Supplier No.:
    06-1101
Spécifications
A propos de cet article
The Pinard Fetoscope, also known as the Pinard Horn, is a type of stethoscope used for listening to the heartbeat of a fetus
There are two types of fetoscopes: the stethoscope for listening to a fetal heartbeat and a fiber-optic scope for viewing the fetus internally. The Pinard Horn is a hollow horn-shaped stethoscope approximately 20.3cm (8") in length, that is pressed against the patient's stomach. The horn must be at a precise location close to the heart in order to hear the fetal heartbeat. Because of this, it is considered to be a more accurate device than the Doppler Fetal Monitor, which picks up tones that can be further from the heart, but has become the fetal heartbeat monitor of choice in the United States. The Pinard Horn is commonly used in Europe by doctors, nurses, and midwives, and is considered to be a more practical, less expensive, and safer alternative to the Doppler device.
Customers Who Bought This Also Bought