ContactUsLinkComponent
DeLee-Hillis Obstetrical Stethoscope with Metal Headband and Rigid Arm, Sklar
DeLee-Hillis Obstetrical Stethoscope with Metal Headband and Rigid Arm, Sklar
# de catalogue 10194-494
Fournisseur:  Sklar
Stethoscope
DeLee-Hillis Obstetrical Stethoscope with Metal Headband and Rigid Arm, Sklar
# de catalogue 10194-494
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Chrome/Black
  • Cat. no.:
    10194-494
  • Length:
    76.2 cm (30")
  • Supplier No.:
    06-1301
Spécifications
A propos de cet article
The DeLee-Hillis Obstetrical Stethoscope, also known as the DeLee-Hillis Fetoscope, features a metal headband attached to a common binaural stethoscope by a rigid metal arm which allows the user to work hands-free while listening to a fetal heartbeat during an examination
The original intent with this design was to allow the obstetrician to be more prepared for impending delivery. Chrome plated/PVC construction.
Customers Who Bought This Also Bought