Waterproof Labels, Wet Grip, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Labels for moist, frosty or frozen surfaces.
- Grips to wet, frost-covered and frozen surfaces; temperature range –80 to100 °C
- Thin material is compatible with centrifuge rotors
- Resistant to chemicals and solvents
- Adheres to plastics, glass, metals and even paperboard boxes
Choose your format: laser sheets for printing many labels at once, direct thermal labels for printing a few labels at a time or labels on a roll for handwriting labels.
Customers Who Bought This Also Bought