Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Grips to wet, frost-covered and frozen surfaces.
- Thin material is compatible with centrifuge rotors
- Resistant to chemicals and solvents
- Adheres to plastics, glass, metals and even paperboard boxes
- Temperature range is –80 to 70 °C
Choose your format: laser sheets for printing many labels at once, direct thermal labels for printing a few labels at a time or labels on a roll for handwriting labels.
Frequently Bought Together