Lables, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers
Quickly label samples with sharp text, clear barcodes or a combination of both. For use in Temperatures From 70 to –196 °C.
Customers Who Bought This Also Bought