Cryo Labels, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers.
Quickly label samples with sharp text, clear barcodes, or a combination of both. For use in temperatures from 70 to –196 °C.
Customers Who Bought This Also Bought