Thermal Transfer Cryogenic Tags, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Thermal transfer Cryo-Tags® withstand long term cryogenic and freezer storage.
Labels are chemically inert with solvent resistant adhesive that adheres to croygenic vials, microtubes, test tubes, and metal racks. Cryo-Tags® withstand flashfreezing in liquid nitrogen, and dry heat in the range of –190 to 80 °C.
Customers Who Bought This Also Bought