Cryo Labels, Laser Printable, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Cryo-Tags® withstand both long term cryogenic storage conditions and conventional freezer storage.
- Withstand flash/snap freezing in liquid nitrogen
- General purpose
- Temperature range between –196 to 79 °C
These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. It can withstand boiling water baths at 100 °C and dry heat up to 150 °C.
Customers Who Bought This Also Bought