Cryo Labels, Laser Printable, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Cryo-Babies® and Cryo-Tags® withstand both long term cryogenic storage conditions and conventional freezer storage.
- Ideal use for 1.5 ml tubes
- Temperature ranges from –196 to 79 °C
These labels are chemically inert with a solvent resistant adhesive that will adhere to cryogenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies® and Cryo-Tags® also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100 °C and dry heat up to 150 °C (see temp. ranges).
Customers Who Bought This Also Bought