Cryo Labels, Laser Printable, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Cryo-Tags® Laser Sidewall Labels can withstand both long term cryogenic storage conditions and conventional freezer storage.
- Assortment includes blue, green, orange, red and yellow colored labels
These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. These labels also can withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150 °C.
Customers Who Bought This Also Bought