Solvent Resistant Tags, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Solvent resistant labels that cleanly remove from surfaces.
- Resistant to most laboratory solvents and have shown short-term resistance to Acetone and Xylene
- When direct thermal printed or mark with a solvent resistant pen, labels exhibit exceptional durability
- Available on rolls for hand writing or for direct thermal printing using the Dymo® LabelWriter
- Temperature resistant from –80 to 93 °C
Great for beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes and easily remove from surfaces without leaving any adhesive.
Customers Who Bought This Also Bought