Breathe-Easy® Sealing Film for Petri Dishes, Diversified Biotech
Catalog # 10141-850
Supplier: Diversified Biotech
Specifications
- L×W:282.58×15.88 mm (11.13×0.63")
- Cat. no.:10141-850
- Supplier No.:BEMP-1000
Specifications
About this item
Strip design allows film to be protected during application; a tab remains attached for easy removal.
- Strips allow for beneficial exchange of oxygen* and CO2 for cellular growth
- Prevent microbial and viral contamination during growth and storage of cells
- Dishes sealed with Breathe Easy strips may be incubated and/or stored at ambient temperatures, refrigerated or frozen
Customers Who Bought This Also Bought