A propos de cet article
The SklarSpec™ Disposable Graves Vaginal Speculum is a clear plastic speculum used for performing pelvic examinations, pap smears, and examining the cervix.
- Provides 360° visibility
- One hand operation
- Greatly reduces temperature shock
Speculum provides one hand operation for the user and greatly reduces the temperature shock commonly experienced by patients when a metal speculum is used. It is a double blade duckbill speculum inserted with the blades closed.
The blades are then opened by an external lever. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Since these are disposable in nature, SklarSpec™ instruments reduce the likelihood of cross-contamination and eliminate sterilization and reprocessing time and costs. These instruments are available with or without a built-in LED light source, sterile or non-sterile, and in multiple sizes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 75817-550
- 75995-700
- 75995-698
- 10192-434
- 10185-246
- 10193-818
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free