Quartz Tubing, Fused, Thermo Scientific
Fournisseur: Thermo Scientific Chemicals
Certificats
A propos de cet article
These are manufactured by fusing naturally occuring crystalline silica.
- High chemical impurity
- High corrosion resistance
- High melting point
- Extreme hardness
- Low coefficient of thermal expansion
- Excellent electrical insulation quality
Optical transmission from ultra-violet to infra-red.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.