Dishes, Evaporating, Glassy Carbon, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
These evaporating dishes are made up of glassy carbon.
- Resistance to all wet decomposition agents
- No memory effects (uncontrolled adsorption and release of foreign elements)
- No contamination of analytical samples
- Stability to acid and alkaline melts
- No wetting effect of metal melts
- Good resistance to thermal shock allows rapid heating and cooling times
- Resistive to abrasive wear
- Good electrical conductivity
⁴/₄ ⁴/₄ -1 -1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.