Clear Parts Cabinets
Fournisseur: Treston
Certificats
A propos de cet article
Cabinets, which combine a stylish industrial design with practical accessories to provide flexible and efficient small parts storage
- Available in two cabinet heights, 21.65" and 11.42"
- Designed with drawer handles recessed within the frame for additoinal protection during transport
- Can be wall mounted, placed on worksurface, or mounted on space-saving turntables
The crystal clear drawers are transparent, allowing visual identification. Each drawer has a corrugated base to simplify picking. A large, sturdy handle provides a full width label area. Optional accessories: width and length dividers.
A compact storage system: one cabinet offers storage for up to 360 different items. Designed with drawer handles recessed within the frame for additional protection during transport. The cabinets can be wall mounted, placed on the worksurface, or mounted on space-saving turntables.
Ordering information: Installation is required. Assembly videos available.
Customers Who Bought This Also Bought