Micro Roll, Microfiber Wipes, Vileda Professional-FHP
Catalogue # 10769-328
Fournisseur: Vileda Professional - FHP
Spécifications
- Color:White
- Cat. no.:10769-328
- Supplier No.:149041
Spécifications
À propos de cet article
A lightweight, true 100% microfiber wipe offering outstanding lint free cleaning performance with greater than 99.93% removal of bacteria.
- 100% endless microfiber offers streak-free performance
- Lint-free cleaning performance with greater than 99.93% bacteria removal
- One step cleaning with no additional workload
- Better cleaning results in less time
- Suitable for bucket or spray cleaning methods
This semi-disposable cloth features one-step cleaning and streak free performance. Achieve better cleaning results in less time, and these wipes are suitable for bucket or spray cleaning methods as well.
Ordering information: Wipes are packaged 200 wipes per roll with 4 rolls per case. Wipes are 50 gm per sheet.