Whatman™ Grade 520 a ½ Qualitative Filter Papers, Whatman products (Cytiva)
Fournisseur: Cytiva
Certificats
A propos de cet article
A thin paper with great wet strength and a very high flow rate. Frequently used in technical applications such as the filtration of viscous liquids and emulsions (e.g., sweetened juices, spirits and syrups, resin solutions, oils, or plant extracts).
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
- Increased total loading capacity as more filter area is available
- Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
- The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.
Also available in flat stock form as Grade 520 a.
Spécifications
- Catalog No:
- CA11013-898
- CA11013-956
- CA76423-748
- Thickness:
- 300 µm
- 300 µm
- 300 µm
- Particle retention:
- 18 µm (in Liquid Max)
- 18 µm (in Liquid Max)
- 18 µm (in Liquid Max)
- Filtration speed:
- Approx. 17.5 s (Herzberg)
- Approx. 17.5 s (Herzberg)
- Approx. 17.5 s (Herzberg)
- Basis weight:
- 90 g/m²
- 90 g/m²
- 90 g/m²