ContactUsLinkComponent
Organic Vapor Filter , Quality Lab Accessories
Organic Vapor Filter , Quality Lab Accessories
Catalog # 89132-996
Supplier:  QLA
Organic Vapor Filter , Quality Lab Accessories
Catalog # 89132-996
Supplier:  QLA
Specifications
  • Cat. no.:
    89132-996
  • Supplier No.:
    EF-101
Specifications
About this item
Organic vapor filter.
Used on SLVSAF-01 and SLVSAF-02 solvent safe HPLC waste collection products.
Customers Who Bought This Also Bought