All-In-One Jacketed Bench Top Reactor, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This jacketed, one liter or two liter general-purpose bench top reactor system is designed so that all wetted surfaces are glass and PTFE for maximum chemical compatibility
Working temperature range: -50 to 200°C. Working pressure range: Atmosphere to 5 torr. Maximum operating limits: 333 RPM, 3.64 in-lbs of torque.
Supplied with all parts necessary for operation. Jacket inlet/outlet has either 28/15 o-ring ball joint connection, or #11 Ace-Thred™ connection.
A small stand footprint and flexible drive shaft combination allow for the reactor system to be placed in almost any laboratory hood configuration
Customers Who Bought This Also Bought