Insulating Blankets, Pilot Plant, Ace Glass Incorporated
Fabric insulating blankets for jacketed pilot plant flasks
Fits between the inlet/outlet ports on the flasks. Has laced front for easy on/off. Non-heating nor electric.
1" thick blanket has a grey PTFE coated fabric outside for chemical resistance with approximately 1" of insulating material internally for excellent insulating of temperature on jacketed flasks.
Customers Who Bought This Also Bought