Digitrol II Temperature Control, Ace Glass Incorporated
# de catalogue 10767-342
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Cat. no.:10767-342
- Electrical:120 V, 50/60 Hz
- Probe Type:J, K, & T
- Supplier No.:12090-22
Spécifications
A propos de cet article
The DigiTrol II temperature controller displays both setpoint and process temperature.
- Automatic temperature control
- Auto-tune feature
Microprocessor-based, this controller features automatic temperature control and an auto-tune feature which minimizes overshoot as it learns your process. It can perform simple ramp rate operations to slowly raise the process temperature and is input sensor type selectable, accepting J, K and T type probes (not included) accommodated by a universal input sensor socket. The unit has a rear panel grid support bracket and a 6 foot detachable power cord.
Certifications: cULus
Ordering information: J, K and T type probes are not included. Inquire if needing a 240 V, CE rated unit; contact your Avantor representative for more information. One year manufacturer’s limited warranty.
Customers Who Bought This Also Bought