Right Angle Tube Compression Adapter with Ground Glass Sidearm, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These glass standard taper and spherical joint adapters feature a right angle ground glass sidearm.
- Glass [ST] or spherical joint
- Right angle sidearm
The sidearm is used to make a tube compression fitting connection to a wide variety of flexible and rigid tubing, using PTFE fittings.
Customers Who Bought This Also Bought