PTFE Tube Compression Union, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The PTFE tube compression unions have body and ferrules made of virgin PTFE
Performance at ambient room conditions: 120psi for 1/16" fittings linear decreasing to 80psi for 3/4" fittings and 60psi for fittings larger than 3/4".
Performance at elevated temperatures up to 85C/185F: 90psi for 1/16" fittings linear decreasing to 60psi for 3/4" fittings and 40psi for fittings larger than 3/4".
The nut and ferrule-gripper are made of PVDF.
Customers Who Bought This Also Bought