Right Angle Tube Compression Adapter, with Drip Tip, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The glass standard taper joint adapters feature a drip tip and a right angle ground glass tube sidearm.
- Glass [ST] joint
- Drip tip
- Right angle sidearm
The sidearm is used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.
Customers Who Bought This Also Bought