Vertical Tube Compression Adapters, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These glass standard taper and spherical joint adapters feature a vertical ground glass sidearm.
- Vertical sidearm
- Glass [ST] or spherical joint
The sidearm is used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.
Customers Who Bought This Also Bought