Right Angle Tee Joint to Tube Compression Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These glass standard taper and spherical joint adapters feature right angle tee ground glass sidearms.
- Glass [ST] or spherical joint
- Right angle tee sidearms
The sidearms are used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.
Customers Who Bought This Also Bought