Jacketed Kilo Scale Reactor, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
A jacketed reactor base system that is designed for maximum diversity and ease of use
Each base system comes with the basic starting components required. The motor and accessories should be purchased separately, based upon your specific needs.
Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom-designed parts.
Ordering information: Base reactor system includes universal stand with motor mount, jacketed flask, multi-neck head, O-ring/gasket and quick-release clamp for head, cap, O-ring, and quick-release clamp for addition port on head, PTFE Ace-Thred bearing for stir shaft, glass stir shaft, shaft collar and shaft coupling, anchor-style agitator, 45° agitator, flush drain seal valve with O-ring and beaded pipe couplings.
Customers Who Bought This Also Bought