Unjacketed Spherical Kilo Scale Reactor Base Systems, Flange Mount, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
An unjacketed spherical reactor base system that is designed for maximum diversity and ease of use
Each base system comes with the basic starting components required.
The motor and accessories should be purchased separately, based upon your specific needs.
Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom-designed parts.
Ordering information: Base reactor system includes universal stand with motor mount, unjacketed spherical flask with flush seal valve, three neck head, CAPFE O-ring and quick-release clamp for head, PTFE Ace-Thred bearing for stir shaft, glass stir shaft, shaft collar and shaft coupling, 45° upper turbine, multi-paddle style lower agitator, flush drain seal valve with O-ring, beaded pipe coupling, aluminum heating mantle, girdle mantle, pilot plant temperature controller, thermocouple, thermocouple extension cord, Ace-Thred adapter for thermocouple, reducing adapter, PTFE-clad copper heat exchange cooling coil, and Ace-Thred adapters for coil.
Customers Who Bought This Also Bought