APS™ Adhesion Microscope Slides, Matsunami Glass
Fournisseur: Matsunami Glass
Certificats
A propos de cet article
The Matsunami APS™ slide is treated to provide a strong permanent charge to the slide surface
Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass. Slides are available with 90° or clipped corners.
Excellent for routine histology, IHC, and cytology preparations. Optimized for use with thermal printers.
Ordering information: Slides are packaged 100 per box, 10 boxes per case (1000 slides).
Customers Who Bought This Also Bought