Human Sperm
# de catalogue 470177-646
Fournisseur: Avantor
Spécifications
- Description:Human Sperm, smear
- Cat. no.:470177-646
- Stain:Kernechtrot Picroindigocarmine
- Supplier No.:WARD470177-646
- Orientation:Smear
Spécifications
A propos de cet article
Smear, Kernechtrot-Picroindigocarmine (KPIC) “Christmas tree” stain.
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Kernechtrot-Picroindigocarmine (KPIC) “Christmas tree” stained slides for greater contrast. Will show spermatozoa red bodies with the midpiece and tail sections green.
Customers Who Bought This Also Bought