Vis-Tech Hi-Vis Cut Resistant Gloves with Sandy Nitrile Palm, Wells Lamont
Fournisseur: Wells Lamont
Certificats
À propos de cet article
Palm coated work gloves provide additional protection in high wear/high risk areas of the hand. The Y9296 is manufactured using a unique blend of high performance fibers and stainless steel to achieve a high level of comfort and cut resistance.
- Sandy nitrile coated palm provides a non-slip surface in wet, dry and oily conditions
- Open, uncoated back circulates air, allowing hands to ‘breathe’ and keeping them cool and comfortable
- Hi-vis cut resistant shell provides a high level of protection & creates awareness in tough environments
- Knit wrist construction prevents dirt and debris from entering the glove
The Y9296 is an excellent solution for applications requiring good oil grip and dexterity along with cut protection.
Lightweight and breathable, the gloves can be placed in the washing machine.
Certifications: ANSI Cut Level 4.
Ordering information: Gloves are sold by the dozen pairs.
Spécifications
- Catalog No:
- 10014-184
- 10014-186
- 10014-188
- 10014-190
- 10014-192
- 10014-194
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Nitrile palm coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist