SklarLite™ Mid Size Rigid Sterilization Container Lids, Sklar
Fournisseur: Sklar
Certificats
A propos de cet article
The SklarLite™ Mid Size Rigid Sterilization Container Lids are available in a variety of colors.
- Available in perforated or solid formats
Made of anodized aluminum alloy, these reusable lids are nonsterile and latex-free.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.