Diskcover-47 Reservoir, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The Diskcover-47 open-top reservoir allows users to pour up to 125 mL of sample directly onto the filter disk holder.
- Easy installation on Diskcover-47 extraction disk holder
Customers Who Bought This Also Bought