Accessories for Resprep® SPE Manifolds, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$46.59 to $465.92
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$137.63
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$180.01 to $182.41
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Fournisseur: MicroSolv Technology Corp
$54.30