Resprep® Sample Delivery System, Restek
# de catalogue 10803-402
Fournisseur: Restek
Spécifications
- Description:Resprep® Sample Delivery System
- Cat. no.:10803-402
- Supplier No.:26250
Spécifications
A propos de cet article
Compatible with Resprep® 1 ml, 3 ml, 6 ml, and 15 ml SPE cartridges and the Diskcover-47 extraction disk holder.
- Colored bands for easy identification
- Compatible with SPE cartridges of different sizes
- Prevents any contaminants leaching from system
The delivery system features six PTFE transfer lines (1/8" O.D.×1/16" I.D.×36" L). Each line is banded with a different color for easy sample identification. Specified in EPA drinking water methods. Tested to pH of 1 to ensure no contaminants leach from system.
Ordering information: Use the Resprep® sample delivery system to transfer large volumes of low-viscosity samples directly from a bottle to a solid phase extraction cartridge, or to a disk on a vacuum manifold system for extraction or cleanup.
Customers Who Bought This Also Bought