Shaker Flask Clamps, Triforest Enterprises
Fournisseur: TRIFOREST ENTERPRISES INC
Certificats
A propos de cet article
Polypropylene shaker clamps are carefully engineered to provide both clipping elasticity and motion resistance in one product
Featuring a non-scratch design, these shaker clamps come in standard blue color accompanied by stainless steel screws and washers. Clamps fit all industry standard shaking incubators and are autoclavable for reuse.
Customers Who Bought This Also Bought