Boekel® Warmer, Bioptechs Inc.®
Fournisseur: BIOPTECHS INC.
Certificats
A propos de cet article
The Boekel® Warmer is a non-humidified, heated enclosure designed for storing items to be warmed adjacent to a microscope
The enclosure measures 20.3 x 20.3 x17.8 cm (8 x 8 x 7"), and is available in 120V or 230V formats.
Certifications: CE certified.
Ordering information: Unit is supplied as heater only.
Customers Who Bought This Also Bought