Objective Wrench and Plier Set, Bioptechs
# de catalogue 10199-798
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:Objective Wrench and Plier Set
- Cat. no.:10199-798
- Supplier No.:021523
Spécifications
A propos de cet article
This tool set used to isolate the mechanical stress applied to an objective when removing the decorative collar.
- Safely allows removal of decorative collar
- For use with the Objective Heater® System
Designed to enable the attachment of an objective heater.
Certifications: CE certified.
Customers Who Bought This Also Bought