FCS3® Spanner Wrench, Bioptechs
# de catalogue 10199-792
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:FCS3® Spanner Wrench
- Cat. no.:10199-792
- Supplier No.:2106031931923
Spécifications
A propos de cet article
A knurled aluminum disk featuring pins inserted to interface the FCS3® chamber
Facilitates easy assembly.
Certifications: CE certified.
Customers Who Bought This Also Bought