FCS2® Non-Heated Flow Cell, Bioptechs®
# de catalogue 10200-010
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:FCS2® Non-Heated Flow Cell
- Cat. no.:10200-010
- Supplier No.:030603192NH
Spécifications
A propos de cet article
A parallel plate, high N.A
- Near laminar flow perfusion
- User definable flow characteristics
- Compatible with all modes of microscopy including high NA/high magnification uses
- Over 20 years of being the foremost chamber in micro-environmental control
- Ideal for both flow and static applications
Compatible, microscopic flow cell with a user definable specimen area.
Certifications: CE certified.
Delivery: Includes Non-Heated FCS2® Chamber, five non-heating micro aqueduct slides, one hundred #1.5 thickness, 40 mm German Desag263 glass Coverslips, and a 30-piece Gasket Set.
Customers Who Bought This Also Bought